Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198914.1 | NP_001185843.1 | 15 | 2381 | ||
NM_010848.3 | NP_034978.3 | 14 | 2024 | |||
GenBank mRNA | AK088020.1 | - | 1 | 54 | ||
AK088079.1 | - | 15 | 2302 | |||
AK132250.1 | - | 14 | 1889 | |||
AK137794.1 | - | 8 | 969 | |||
AK137965.1 | - | 14 | 1890 | |||
BC011513.1 | - | 14 | 1932 | 3672769 | ||
M12848.1 | - | 14 | 2022 | |||
X02774.1 | - | 14 | 1794 |