Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010851.2 | NP_034981.1 | 5 | 841 | ||
GenBank mRNA | AK150083.1 | - | 5 | 724 | ||
AK151748.1 | - | 5 | 797 | |||
AK152596.1 | - | 5 | 826 | |||
AK152935.1 | - | 5 | 799 | |||
BC005591.1 | - | 5 | 841 | 3482246 | ||
BC051075.1 | - | 5 | 788 | |||
BC058787.1 | - | 5 | 776 | 6309465 | ||
DQ790885.1 | - | 5 | 760 | |||
DQ790886.1 | - | 5 | 760 | |||
U84409.1 | - | 5 | 822 | |||
X51397.1 | - | 5 | 710 |