Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177369.3 | NP_796343.2 | 20 | 2454 | ||
GenBank mRNA | AK029328.1 | - | 20 | 2365 | ||
BC150737.1 | - | 20 | 2377 | 9087648 |