Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164171.1 | NP_001157643.1 | 7 | 824 | ||
NM_010856.4 | NP_034986.1 | 7 | 760 | |||
GenBank mRNA | AK142232.1 | - | 7 | 797 | ||
BC061145.1 | - | 7 | 793 | |||
BC090979.1 | - | 7 | 793 | |||
BC110700.1 | - | 7 | 806 | 6786621 | ||
BC138027.1 | - | 6 | 699 | 8861046 | ||
M76598.1 | - | 5 | 622 | |||
M76599.1 | - | 5 | 622 | |||
M76600.1 | - | 5 | 622 | |||
M76601.1 | - | 7 | 724 |