Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010858.4 | NP_034988.2 | 5 | 330 | ||
GenBank mRNA | AK013098.1 | - | 3 | 258 | ||
AK084544.1 | - | 1 | 1968 | |||
BC048485.1 | - | 5 | 330 | 6741975 | ||
M19436.1 | - | 1 | 12 |