Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010859.2 | NP_034989.1 | 4 | 416 | ||
GenBank mRNA | AK002312.1 | - | 4 | 418 | ||
AK011518.1 | - | 4 | 409 | |||
AK146943.1 | - | 4 | 420 | |||
BC061222.1 | - | 4 | 419 | 30297023 |