Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022879.2 | NP_075017.2 | 3, 4 | 221 | ||
GenBank mRNA | AJ348866.1 | - | 2, 3 | 131 | ||
AK146716.1 | - | 3, 4 | 222 | |||
BC061100.1 | - | 3, 4 | 217 | 30300846 | ||
CA461005.1 | - | 3, 4 | 258 | |||
S70785.1 | - | 2, 3 | 166 |