Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010861.3 | NP_034991.3 | 4 | 228 | ||
GenBank mRNA | AK002367.1 | - | 4 | 229 | ||
AK146674.1 | - | 4 | 232 | |||
BC061144.1 | - | 229 | 30301284 | |||
M91602.1 | - | 4 | 220 |