Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016754.5 | NP_058034.1 | 5 | 496 | ||
GenBank mRNA | AK003149.1 | - | 5 | 401 | ||
AK010483.1 | - | 5 | 497 | |||
BC055869.1 | - | 5 | 380 | 4018694 | ||
U77943.1 | - | 4 | 343 |