Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053214.2 | NP_444444.2 | 12 | 1360 | ||
GenBank mRNA | AK036840.1 | - | 2 | 1193 | ||
AK131188.1 | - | 2 | 53 | |||
AK153908.1 | - | 12 | 1323 | |||
AK158333.1 | - | 12 | 1275 | |||
BC046502.1 | - | 12 | 1348 | 4167558 | ||
X97650.1 | - | 12 | 1278 |