Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201600.2 | NP_963894.1 | 27 | 3735 | ||
GenBank mRNA | AK173102.1 | - | 4 | 415 | ||
BC011494.1 | - | 6 | 694 | |||
BC046444.1 | - | 18 | 2444 | |||
BC118525.1 | - | 27 | 3728 | 40099275 | ||
BU610647.1 | - | 2 | 106 | |||
BY245557.1 | - | 4 | 329 | |||
M55253.1 | - | 3 | 370 |