Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256081.1 | NP_001243010.1 | 14 | 1934 | ||
NM_001256082.1 | NP_001243011.1 | 14 | 1781 | |||
NM_001256083.1 | NP_001243012.1 | 13 | 1763 | |||
NM_008663.2 | NP_032689.2 | 14 | 1934 | |||
GenBank mRNA | AY821853.1 | - | 13 | 1770 | ||
AY821854.1 | - | 14 | 1788 |