Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032394.3 | NP_115770.2 | 47 | 6375 | ||
GenBank mRNA | AF242411.1 | - | 47 | 6412 | ||
AK220152.1 | - | 22 | 2956 | |||
BC026416.1 | - | 20 | 2719 | |||
BC138338.1 | - | 47 | 6375 | 8861358 | ||
BC138341.1 | - | 47 | 6375 | 8861361 |