Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010865.3 | NP_034995.3 | 1 | 178 | ||
GenBank mRNA | AB013592.1 | - | 1 | 162 | ||
AF039869.1 | - | 1 | 159 | |||
AK144395.1 | - | 1 | 174 | |||
BC125329.1 | - | 1 | 123 | 40129844 | ||
BC137606.1 | - | 1 | 123 | 8860625 | ||
BY337559.1 | - | 1 | 178 |