Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027892.2 | NP_082168.1 | 10 | 1438 | ||
GenBank mRNA | AK004785.1 | - | 10 | 1438 | ||
AK030460.1 | - | 1 | 40 | |||
AK035230.1 | - | 2 | 225 | |||
AK139570.1 | - | 10 | 1324 | |||
BC125381.1 | - | 11 | 1507 | 40129896 | ||
BC137630.1 | - | 11 | 1507 | 8860649 |