Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008667.3 | NP_032693.2 | 3 | 1169 | ||
GenBank mRNA | AB221677.1 | - | 2 | 911 | ||
BC005627.1 | - | 3 | 1169 | 3155795 | ||
BC016886.1 | - | 3 | 1092 | 4236565 | ||
U47008.1 | - | 3 | 1047 |