Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122895.1 | NP_001116367.1 | 2 | 1052 | ||
NM_008668.2 | NP_032694.2 | 2 | 1052 | |||
GenBank mRNA | AK155501.1 | - | 2 | 950 | ||
AK158688.1 | - | 2 | 982 | |||
AK171180.1 | - | 2 | 932 | |||
BC045139.1 | - | 2 | 936 | 6306757 | ||
U47543.1 | - | 2 | 1027 |