Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008669.4 | NP_032695.3 | 4 | 446 | ||
GenBank mRNA | AF079458.1 | - | 4 | 404 | ||
AK077116.1 | - | 4 | 418 | |||
AK133576.1 | - | 4 | 404 | |||
AK153002.1 | - | 4 | 432 | |||
BC021631.1 | - | 4 | 426 | 4019197 | ||
CJ093426.1 | - | 4 | 449 |