Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010871.2 | NP_035001.2 | 14 | 5150 | ||
GenBank mRNA | AF135494.1 | - | 5274 | |||
AK136436.1 | - | 6 | 2412 |