Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008673.1 | NP_032699.1 | 1 | 965 | ||
GenBank mRNA | BC119181.1 | - | 1 | 770 | 8733930 | |
BC137747.1 | - | 1 | 770 | 8860766 | ||
U35885.1 | - | 1 | 965 |