Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168577.1 | NP_001162049.1 | 2 | 594 | ||
NM_010874.3 | NP_035004.1 | 2 | 692 | |||
GenBank mRNA | AJ251710.1 | - | 2 | 692 | ||
AK007583.1 | - | 2 | 603 | |||
U35886.1 | - | 1 | 776 | |||
U35887.1 | - | 1 | 776 |