Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252220.1 | NP_001239149.1 | 11 | 1422 | ||
NM_001252222.1 | NP_001239151.1 | 11 | 1245 | |||
NM_001252223.1 | NP_001239152.1 | 11 | 1245 | |||
NM_008676.3 | NP_032702.1 | 11 | 1245 | |||
GenBank mRNA | AF227188.1 | - | 11 | 1374 | ||
AF408436.1 | - | 2 | 83 | |||
AK077479.1 | - | 1 | 30 | |||
AK128992.1 | - | 11 | 1037 | |||
AK129043.1 | - | 11 | 1015 | |||
AK154081.1 | - | 11 | 1031 | |||
BC052709.1 | - | 11 | 1035 | 6417055 | ||
BC129867.1 | - | 10 | 897 | 40110124 | ||
BC129868.1 | - | 10 | 897 | 40110128 | ||
U73039.1 | - | 11 | 1041 |