Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008677.2 | NP_032703.2 | 7 | 807 | ||
GenBank mRNA | AB002665.1 | - | 7 | 784 | ||
AK150510.1 | - | 7 | 789 | |||
AK170854.1 | - | 4 | 315 | |||
AK171315.1 | - | 7 | 585 | |||
BC025517.1 | - | 7 | 636 | 5321182 | ||
FJ168470.1 | - | 7 | 598 | |||
U59488.1 | - | 7 | 754 |