Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010881.2 | NP_035011.1 | 5 | 940 | ||
GenBank mRNA | BC068177.1 | - | 2 | 248 | 30628581 | |
BC080866.1 | - | 5 | 703 | 30532122 | ||
U56920.1 | - | 4 | 498 | |||
U64606.1 | - | 2 | 243 | |||
U64828.1 | - | 5 | 853 |