Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB219976.1 | - | 9 | 1386 | ||
AF000582.1 | - | 11 | 1537 | |||
AK052117.1 | - | 11 | 1608 | |||
AK087443.1 | - | 12 | 1616 | |||
BC053387.1 | - | 11 | 1731 | 30054358 | ||
U39060.1 | - | 11 | 1589 |