Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010890.3 | NP_035020.2 | 23 | 2315 | ||
GenBank mRNA | AK088620.1 | - | 23 | 2219 | ||
AK088767.1 | - | 23 | 2228 | |||
AK122203.1 | - | 23 | 2158 | |||
AK133838.1 | - | 23 | 2228 | |||
AK144240.1 | - | 23 | 2228 | |||
BC007184.1 | - | 3 | 162 | |||
BC138813.1 | - | 23 | 2193 | 8861835 | ||
D85414.1 | - | 23 | 2315 | |||
U96635.1 | - | 23 | 2386 |