Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111324.2 | NP_001104794.1 | 6 | 1841 | ||
NM_017464.5 | NP_059492.3 | 5 | 1622 | |||
GenBank mRNA | AF009366.1 | - | 6 | 1777 | ||
AK030985.1 | - | 6 | 1775 | |||
AK033729.1 | - | 5 | 1623 | |||
AK046357.1 | - | 5 | 1585 | |||
AK054179.1 | - | 5 | 1618 | |||
AK083374.1 | - | 6 | 1769 | |||
BC053713.1 | - | 6 | 1774 | 30077589 |