Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010893.3 | NP_035023.3 | 4 | 903 | ||
GenBank mRNA | AK077444.1 | - | 3 | 2018 | ||
AK145680.1 | - | 4 | 797 | |||
AK190751.1 | - | 1 | 27 | |||
AK200961.1 | - | 1 | 27 | |||
BC004666.1 | - | 4 | 777 | 3498197 | ||
U93702.1 | - | 4 | 779 | |||
Y11412.1 | - | 4 | 782 |