Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010896.2 | NP_035026.1 | 1 | 1360 | ||
GenBank mRNA | BC062148.1 | - | 1 | 1360 | 30146192 |