Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122952.1 | NP_001116424.1 | 2 | 446 | ||
NM_001122953.1 | NP_001116425.1 | 2 | 685 | |||
NM_010905.3 | NP_035035.1 | 2 | 685 | |||
GenBank mRNA | AF326553.1 | - | 2 | 384 | ||
AF326554.1 | - | 2 | 384 | |||
AK052204.1 | - | 2 | 507 | |||
AK054310.1 | - | 2 | 549 | |||
AK137731.1 | - | 2 | 548 | |||
AK173173.1 | - | 1 | 321 | |||
BC075702.1 | - | 2 | 666 | 30087137 | ||
BC122878.1 | - | 2 | 407 | 40107695 | ||
BC122880.1 | - | 2 | 405 | |||
D90172.1 | - | 2 | 615 | |||
D90173.1 | - | 2 | 523 | |||
D90174.1 | - | 2 | 556 | |||
D90175.1 | - | 400 | ||||
D90176.1 | - | 2 | 438 | |||
U57633.1 | - | 2 | 340 | |||
Y07690.1 | - | 2 | 416 | |||
Y07691.1 | - | 2 | 414 |