Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008688.3 | NP_032714.1 | 2 | 270 | ||
NM_026756.2 | NP_081032.2 | 2 | 270 | |||
GenBank mRNA | AK087539.1 | - | 2 | 255 | ||
AK147496.1 | - | 2 | 255 | |||
AK155258.1 | - | 2 | 272 | |||
U57635.1 | - | 2 | 210 | |||
Y07692.1 | - | 2 | 229 | |||
Y07693.1 | - | 2 | 575 |