Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017373.3 | NP_059069.1 | 2 | 759 | ||
GenBank mRNA | AK087541.1 | - | 2 | 763 | ||
AK145451.1 | - | 2 | 769 | |||
AY061760.1 | - | 1 | 431 | |||
BB872092.1 | - | 1 | 360 | |||
BC031377.1 | - | 2 | 603 | 5253195 | ||
BQ899715.1 | - | 1 | 137 |