Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008689.2 | NP_032715.2 | 12 | 1521 | ||
GenBank mRNA | AB119195.1 | - | 10 | 1019 | ||
AB221614.1 | - | 10 | 1026 | |||
AK052726.1 | - | 7 | 829 | |||
AY521462.1 | - | 10 | 1019 | |||
AY521463.1 | - | 10 | 1019 | |||
BC050841.1 | - | 12 | 1521 | |||
BC138535.1 | - | 11 | 1098 | 8861555 | ||
BC138536.1 | - | 11 | 1098 | 8861556 | ||
BC145317.1 | - | 12 | 1115 | |||
M57999.1 | - | 12 | 1310 |