Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177369.1 | NP_001170840.1 | 5, 6 | 403 | ||
NM_001177370.1 | NP_001170841.1 | 4, 5 | 319 | |||
NM_019408.3 | NP_062281.1 | 4, 5 | 554 | |||
GenBank mRNA | AF155372.1 | - | 5, 6 | 481 | ||
AF155373.1 | - | 5, 6 | 518 | |||
AK134081.1 | - | 5, 6 | 822 | |||
AK137680.1 | - | 5, 6 | 393 | |||
AK144994.1 | - | 5, 6 | 396 | |||
AK145312.1 | - | 4, 5 | 520 | |||
AK148160.1 | - | 5, 6 | 391 | |||
AK150012.1 | - | 5, 6 | 382 | |||
BC027423.1 | - | 4, 5 | 320 | 3672913 | ||
CJ134893.1 | - | 4, 5 | 551 | |||
DQ020182.1 | - | 4, 5 | 251 | |||
DQ020183.1 | - | 3, 4 | 372 | |||
DQ020184.1 | - | 3, 4 | 372 | |||
DQ020185.1 | - | 4, 5 | 251 |