Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010218.1 | - | 3 | 652 | ||
AK038631.1 | - | 3 | 700 | |||
AK156731.1 | - | 3 | 626 | |||
AY176058.1 | - | 3 | 659 | |||
AY176059.1 | - | 3 | 690 | |||
BC021938.1 | - | 3 | 657 | 5136809 | ||
U19799.1 | - | 3 | 671 |