Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110832.1 | NP_001104302.1 | 10 | 1267 | ||
NM_010913.2 | NP_035043.1 | 9 | 1180 | |||
GenBank mRNA | AK004729.1 | - | 10 | 1213 | ||
AK143723.1 | - | 9 | 1142 | |||
AK156487.1 | - | 10 | 1097 | |||
BC029695.1 | - | 9 | 1068 | 5374597 | ||
BC057099.1 | - | 10 | 1169 | 5716111 | ||
M86214.1 | - | 8 | 899 | |||
M86215.1 | - | 9 | 1001 | |||
X55315.1 | - | 9 | 1090 |