Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010914.2 | NP_035044.1 | 4 | 334 | ||
GenBank mRNA | AK010762.1 | - | 4 | 314 | ||
AK011792.1 | - | 4 | 253 | |||
AK079209.1 | - | 4 | 289 | |||
AK146168.1 | - | 4 | 336 | |||
AK154749.1 | - | 4 | 296 | |||
AK155749.1 | - | 4 | 296 | |||
BB858171.1 | - | 4 | 340 | |||
BC010719.1 | - | 4 | 275 | 2811674 | ||
X55316.1 | - | 4 | 330 |