Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048168.2 | NP_001041633.1 | 8 | 896 | ||
NM_001277095.1 | NP_001264024.1 | 7 | 855 | |||
NM_008692.4 | NP_032718.2 | 8 | 969 | |||
GenBank mRNA | AB006181.1 | - | 7 | 749 | ||
AK011327.1 | - | 8 | 925 | |||
AK156353.1 | - | 7 | 709 | |||
AK186792.1 | - | 1 | 31 | |||
AK216147.1 | - | 1 | 31 | |||
AK219678.1 | - | 1 | 31 | |||
BC020117.1 | - | 8 | 895 | 4009737 | ||
BC053723.1 | - | 8 | 899 | 30053020 | ||
BC085261.1 | - | 8 | 895 | 6397760 | ||
BM461330.1 | - | 6 | 600 | |||
U62297.1 | - | 7 | 749 |