Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010916.2 | NP_035046.1 | 2 | 1789 | ||
GenBank mRNA | BC051018.1 | - | 2 | 1765 | 6312358 | |
BC054760.1 | - | 2 | 1766 | |||
M82874.1 | - | 2 | 1753 |