Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000871.2 | NP_000862.1 | 1 | 677 | ||
GenBank mRNA | AY429105.1 | - | 1 | 210 | ||
BC074995.2 | - | 1 | 244 | 30915396 | ||
BC074996.2 | - | 1 | 244 | 30915608 | ||
L41147.1 | - | 1 | 677 |
Set Membership: |
G-protein coupled receptor |