Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013610.2 | NP_038638.1 | 2 | 120 | ||
GenBank mRNA | AK077602.1 | - | 2 | 139 | ||
AK155741.1 | - | 2 | 130 | |||
AK159111.1 | - | 2 | 140 | |||
AK160045.1 | - | 2 | 127 | |||
AK171973.1 | - | 2 | 139 | |||
BC049600.1 | - | 2 | 107 | |||
BC118937.1 | - | 2 | 120 | 40091105 | ||
U91513.1 | - | 2 | 120 |