Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008704.2 | NP_032730.1 | 5 | 836 | ||
GenBank mRNA | AF033377.1 | - | 5 | 571 | ||
AK034899.1 | - | 575 | ||||
AK136353.1 | - | 5 | 651 | |||
AK186726.1 | - | 2 | 244 | |||
BC005629.1 | - | 5 | 575 | 3964056 | ||
M35970.1 | - | 5 | 573 | |||
U85511.1 | - | 5 | 623 |
Gene Symbol: Samhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149865.1 | - | 1196 |