Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077529.2 | NP_001070997.1 | 4 | 578 | ||
NM_008705.5 | NP_032731.1 | 5 | 850 | |||
GenBank mRNA | AK012447.1 | - | 5 | 507 | ||
AK150419.1 | - | 5 | 512 | |||
AK151262.1 | - | 4 | 467 | |||
AK169120.1 | - | 5 | 472 | |||
AK216511.1 | - | 3 | 206 | |||
BC066995.1 | - | 5 | 850 | 5691909 | ||
BC086892.1 | - | 4 | 483 | 6752803 | ||
BC086893.1 | - | 5 | 475 | 6758871 | ||
EF437324.1 | - | 4 | 398 | |||
X68193.1 | - | 4 | 434 |