Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008706.5 | NP_032732.3 | 3 | 353 | ||
GenBank mRNA | AK075568.1 | - | 3 | 354 | ||
S75951.1 | - | 3 | 427 | |||
U12961.1 | - | 3 | 328 |