Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163239.1 | NP_001156711.1 | 6 | 716 | ||
NM_001163241.1 | NP_001156713.1 | 7 | 804 | |||
NM_001163242.1 | NP_001156714.1 | 7 | 733 | |||
NM_020282.3 | NP_064678.1 | 8 | 886 | |||
GenBank mRNA | AF252260.1 | - | 7 | 732 | ||
AK008138.1 | - | 6 | 688 | |||
AK008465.1 | - | 6 | 691 | |||
AK010842.1 | - | 7 | 776 | |||
BC027629.1 | - | 8 | 794 | 1225122 |