Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010925.2 | NP_035055.2 | 8 | 744 | ||
GenBank mRNA | AF312394.1 | - | 7 | 507 | ||
AK077540.1 | - | 8 | 669 | |||
BC022910.1 | - | 7 | 610 | 4165783 | ||
BC065993.1 | - | 8 | 706 | 5684672 | ||
BC065994.1 | - | 8 | 664 | 5685269 | ||
U79773.1 | - | 8 | 696 | |||
U79774.1 | - | 8 | 696 |