Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008710.3 | NP_032736.2 | 359 | |||
GenBank mRNA | AK033282.1 | - | 3 | 300 | ||
AK040460.1 | - | 3 | 320 | |||
AK050478.1 | - | 2 | 264 | |||
AK052061.1 | - | 3 | 298 | |||
AK052234.1 | - | 3 | 344 | |||
AK054332.1 | - | 3 | 345 | |||
AK083413.1 | - | 3 | 345 | |||
AK084546.1 | - | 3 | 342 | |||
AK084604.1 | - | 3 | 342 | |||
AK085260.1 | - | 1 | 1753 | |||
AK085903.1 | - | 3 | 345 | |||
AK087064.1 | - | 3 | 341 | |||
AK088044.1 | - | 3 | 341 | |||
AK090115.1 | - | 362 | ||||
AK129006.1 | - | 3 | 336 | |||
AK159694.1 | - | 3 | 345 | |||
AK168736.1 | - | 2 | 142 | |||
BC008518.1 | - | 3 | 322 | 3601702 | ||
Z49204.1 | - | 2 | 266 |