Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010926.5 | NP_035056.1 | 4 | 552 | ||
GenBank mRNA | AF052621.1 | - | 4 | 554 | ||
AK042704.1 | - | 4 | 420 | |||
AK076710.1 | - | 6 | 1421 | |||
AK143131.1 | - | 4 | 513 | |||
AK148190.1 | - | 4 | 1063 | |||
AK161438.1 | - | 4 | 528 | |||
AK164130.1 | - | 4 | 553 | |||
BC009103.1 | - | 4 | 508 | 3482942 |