Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002160.3 | NP_002151.2 | 25 | 6517 | ||
XM_005251972.3 | XP_005252029.1 | 24 | 6194 | |||
XM_005251973.3 | XP_005252030.1 | 21 | 5375 | |||
XM_005251974.3 | XP_005252031.1 | 19 | 4829 | |||
XM_005251975.3 | XP_005252032.1 | 18 | 4556 | |||
XM_006717096.3 | XP_006717159.1 | 26 | 6743 | |||
XM_006717097.3 | XP_006717160.1 | 24 | 6194 | |||
XM_006717098.3 | XP_006717161.1 | 23 | 5921 | |||
XM_006717101.3 | XP_006717164.1 | 19 | 4829 | |||
XM_011518625.2 | XP_011516927.1 | 23 | 5921 | |||
XM_011518626.2 | XP_011516928.1 | 22 | 5651 | |||
XM_011518628.2 | XP_011516930.1 | 20 | 5102 | |||
XM_011518629.2 | XP_011516931.1 | 20 | 5102 | |||
XM_017014678.1 | XP_016870167.1 | 27 | 7016 | |||
XM_017014679.1 | XP_016870168.1 | 26 | 6743 | |||
XM_017014680.1 | XP_016870169.1 | 26 | 6740 | |||
XM_017014681.1 | XP_016870170.1 | 23 | 5924 | |||
GenBank mRNA | AB210038.1 | - | 25 | 6393 | ||
BC151843.1 | - | 22 | 5566 | 8860362 | ||
GU473235.1 | - | 20 | 5016 | |||
M24630.1 | - | 23 | 4815 | |||
M55618.1 | - | 24 | 6240 | |||
X56160.1 | - | 24 | 6156 | |||
X78565.1 | - | 25 | 6418 |