Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008714.3 | NP_032740.3 | 30 | 5858 | ||
GenBank mRNA | AB101634.1 | - | 3 | 513 | ||
AF508809.1 | - | 30 | 5673 | |||
AK054089.1 | - | 5 | 930 | |||
AK090118.1 | - | 30 | 5833 | |||
AK154528.1 | - | 6 | 1480 | |||
AK157475.1 | - | 5 | 697 | |||
BC138441.1 | - | 30 | 5673 | 8861461 | ||
BC138442.1 | - | 30 | 5673 | 8861462 | ||
Z11886.1 | - | 30 | 5672 | |||
Z21925.1 | - | 1 | 173 |